missing translation for 'onlineSavingsMsg'
Learn More
Isatin, 98%, Thermo Scientific Chemicals Product Code.: 10143010

Isatin, 98%, Thermo Scientific Chemicals

Product Code. 10143010
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10143010

Brand: Thermo Scientific Acros 151491000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-56-5
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00005718
InChI Key JXDYKVIHCLTXOP-UHFFFAOYSA-N
Synonym isatin, indoline-2,3-dione, 2,3-indolinedione, indole-2,3-dione, isatine, 2,3-dioxoindoline, pseudoisatin, tribulin, isotin, isatic acid lactamShow MoreShow Less
PubChem CID 7054
ChEBI CHEBI:27539
IUPAC Name 1H-indole-2,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(=O)N2

Specifications

Melting Point 201°C to 204°C
Color Orange
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 21, 432
Merck Index 15, 5148
Solubility Information Solubility in water: 1.9g/L. Other solubilities: soluble in alkali hydroxide solutions,soluble in ethanol,diethyl ether,benzene
Formula Weight 147.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Isatin, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0428531 Certificate TypeCertificate of Analysis Date10/10/2023 Catalog Number151495000,151491000
Lot NumberA0448679 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number151495000,151491000
Lot NumberA0401675 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number151495000,151491000
Lot NumberA0414387 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number151495000,151491000
Lot NumberA0413603 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number151495000,151491000
Product Identifier
  • Isatin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isatin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.