missing translation for 'onlineSavingsMsg'
Learn More

Hopen™ Iso Air Cleanroom Masks with Ties
GREENER_CHOICE

ISO Air cleanroom masks have long ties for easy donning and are made with the Mass Balance approach from recycled materials.

Brand:  Hopen™ 60.720L

 View more versions of this product

Product Code. 17724789

  • 115.00 € / Pack of 300
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Low particle release: Helmke drum tested for use in clean rooms
  • High filtration efficiency of particles, bacteria and viruses
  • Adaptable to users and protocols with two sizes available
  • Headloops improve comfort for the user by making it easier to put on
  • Softex technology limits risk of particulate contamination while reducing risk of skin irritation for the user
  • Adjustable and malleable nose clip prevents glasses from fogging up
  • ISO AIR range is manufactured in an ISO 8 classified clean room
  • Made in France
Specifications

Specifications

Cleanroom Face Mask
Ties
Pleated
Class 100 (ISO 5)
Industry
White
Large
Grade B
>99%
60.720
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hopen™ Iso Air Cleanroom Masks with Ties > Iso Air Cleanroom Masks with Ties; Color: White; Size: Large; Style: Pleated; Cleanroom Class: Class 100/ISO 5; For Use With (Application): Industry

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.