missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck Product Code.: 15634914
Add to basket

DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck
BIOPHARMA

Product Code. 15634914 Shop All DuPont Products
Small
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy

Product Code. 15634914

Brand: DuPont™ D15466113

Add to basket
Add to basket
This item is not returnable. View return policy

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and exceptionally low linting.

  • Available and in sizes S to XXXL.
  • Clean-processed and gamma-sterilized.
  • Bound internal seams.
  • Tunneled elastication at wrists.
  • Front snap closure for easy donning and doffing.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Controlled Environments Lab Coat
Size Small
Material Tyvek
Color White
Closure Type Snaps
Cuff Style Tunnelized Elastic
Disposable Single-use
Sterility Sterile
Collar Type Bound
Seam Type Bound internal seams
Size (Metric) Chest 88 to 97 cm
Certifications/Compliance Clean-processed and sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1), Full traceability on all sterilized apparel with certificates of sterility available, Suitable for use in GMP class A/B (ISO Class 5) clean rooms, PPE Category I
For Use With (Application) Cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
Show More Show Less
  • Dupont Tyvek - Packaging for CE certified IsoClean garments
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Dupont Tyvek Packaging for CE certified IsoClean garments

    • Dupont Tyvek - IsoClean design
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Dupont Tyvek IsoClean design

      • DuPont Tyvek - IsoClean
        Video Player is loading.
        Current Time 0:00
        Duration 0:00
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:00
         
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected

          DuPont Tyvek IsoClean

        Brochures

        Posters

        Product Information

        Certificates

        A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

        5 results found
        Lot Number Certificate Type Date Catalog Number
        Lot NumberAHS2403176 Certificate TypeCertificate of Sterility Date05/04/2024 Catalog NumberD15466113
        Lot NumberAHS2409140 Certificate TypeCertificate of Sterility Date05/04/2024 Catalog NumberD15466113
        Lot NumberAHS2401165 Certificate TypeCertificate of Sterility Date24/01/2024 Catalog NumberD15466113
        Lot NumberAHS2350165 Certificate TypeCertificate of Sterility Date18/01/2024 Catalog NumberD15466113
        Lot NumberAHS2344133 Certificate TypeCertificate of Sterility Date24/11/2023 Catalog NumberD15466113

        The user should make the risk assessment to determine the protection required.

        Product Content Correction

        Your input is important to us. Please complete this form to provide feedback related to the content on this product.

        Product Title
        DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck >

        By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

        Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.