Learn More
Phagogene™ Ethanol Spray
PHAGO'SPRAY DM is a professional disinfectant cleaner for the disinfection of surfaces in high-risk environments. PHAGO'SPRAY DM can be used as a spray on walls, ceilings and furniture in care and surgery departments.
Brand: Phagogene™ 60416
Description
- Ready to use
- Broad spectrum disinfectant Efficacy on Rotavirus in 5 minutes
- A nozzle with two positions (spray and stop pictograms)
- A removable ergo for use in foam or spray
Specifications
Disinfectant Cleaner | |
Fresh Scent | |
6/Pk. | |
750 mL |
Clear Liquid | |
Non-sterile | |
750 mL |
Safety and Handling
- PHAGOSPRAY DM
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.