missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Kevlar™ Yellow Fabric Sleeves with Thumb Slot
Shop All Ansell Products
Click to view available options
Length (Metric):
254 mm
356 mm
457 mm
Description
- Constructed from high-tech 100% Kevlar™ Aramid fibers para-aramid short fibers.
- Exceptional cut resistance with dexterity.
- The 100% Dupont Kevlar™ Aramid fibers construction offers a very high cut resistance to weight ratio.
- Most sleeves feature thumb slots for protecting the wrist area.

Specifications
Specifications
Product Type | Cut-resistant Sleeve |
Length (Metric) | 356 mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell™ Kevlar™ Yellow Fabric Sleeves with Thumb Slot >
Spot an opportunity for improvement?Share a Content Correction