Learn More
Thermo Scientific™ KingFisher Apex Accessories
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.
Brand: Thermo Scientific™ 24079940
Description
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality
- User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
- Provides versatile and high-speed purification with reproducible results
- Easy to use interface with touch screen empowering the user to control all details of the application and purification process
- BindIx PC Software for added convenience
- Extensive selection of validated protocols for different type of applications
- Cloud library for quick protocol retrieval
Specifications
24 Combi Head | |
24 | |
KingFisher Apex | |
ca. >1 μm | |
24 DW, 24 storage tube |
24 DW plate 30 to 5000 μL, 24 storage tube 200 to 1000 μL | |
24 Samples | |
24 | |
24 |
Product Suggestions
Customers who viewed this item also viewed.