missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
18466738
|
Thermo Scientific™
24075970 |
Matrix Heating Block |
700.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16552442
|
Thermo Scientific™
24079910 |
96 PCR Head |
6010.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16562442
|
Thermo Scientific™
24079920 |
Combi Head |
5970.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16572442
|
Thermo Scientific™
24079930 |
96 DW Head |
6000.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16582442
|
Thermo Scientific™
24079940 |
24 Combi Head |
5980.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16592432
|
Thermo Scientific™
24075910 |
PCR Plate Heating Block |
698.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16502442
|
Thermo Scientific™
24075920 |
96 KF Heating Block |
694.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16512442
|
Thermo Scientific™
24075930 |
96 DW Heating Block |
694.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16522442
|
Thermo Scientific™
24075940 |
24 DW Heating Block |
688.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16532442
|
Thermo Scientific™
24075950 |
Heating Block 96 |
684.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16542442
|
Thermo Scientific™
24075960 |
Heating Block 24 |
678.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16572452
|
Thermo Scientific™
N21445 |
Storage Tube Adapter |
476.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16582452
|
Thermo Scientific™
N21446 |
Plate Adapter |
476.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16592452
|
Thermo Scientific™
N21447 |
UV Lamp (Replacement) |
189.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality
- User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
- Provides versatile and high-speed purification with reproducible results
- Easy to use interface with touch screen empowering the user to control all details of the application and purification process
- BindIx PC Software for added convenience
- Extensive selection of validated protocols for different type of applications
- Cloud library for quick protocol retrieval
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ KingFisher Apex Accessories