missing translation for 'onlineSavingsMsg'
Learn More
Ansell KleenGuard™ A40 Liquid and Particle Protection Aprons Product Code.: 12631486

Ansell KleenGuard™ A40 Liquid and Particle Protection Aprons
GREENER_CHOICE

Product Code. 12631486
Universal
Click to view available options
Size:
Universal
This item is not returnable. View return policy

Product Code. 12631486

Brand: Ansell 44481

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

KleenGuard™ A40 Liquid and Particle Protection Aprons feature a breathable, microporous film laminate that protects against nonhazardous liquids.

  • White, disposable aprons
  • Ties in the back
  • Breathable, microporous film laminate
  • Excellent liquid and particulate barrier
  • Low in lint

Compliance

Meet NFPA™ 99 for Antistatic Materials and ASTM™ F1670 testing for penetration of blood and bodily fluids.

Greener Choice Claims
  • Environmental benefits include:
    • Recyclable

Specifications

Type Apron
Size Universal
Material 23-P Film Laminate
Color White
Disposable Reusable
Closure Type Ties
Sterility Non-sterile
For Use With (Application) Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance, general manufacturing
Certifications/Compliance NFPA 99, ASTM F1670
Dimensions (L x W) 101.6 x 71 cm (40 x 28 in.)
Length (English) 40 in.
Length (Metric) 101.6 cm
Width (English) 28 in.
Width (Metric) 71 cm
Item Description Kleenguard A40 Light Duty Short Apron - Apron / White
Seam Type Sewn
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell KleenGuard™ A40 Liquid and Particle Protection Aprons >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.