Learn More
DWK Life Sciences Kimble™ Kontes™ Microcaps™ TLC Spotting Capillaries
For spotting preadsorbent TLC plates
Brand:  DWK Life Sciences 764520-0000
427.84 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 13075813
  • 539.00 € / Pack of 500
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Kit includes:
  • Five dispenser vials containing 100 micropipets each of 0.5, 1, 2, 5 and 10μL
  • Bulb assembly
  • Spotting holder
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
Five dispenser vials containing 100 micropipets each of 0.5, 1, 2, 5 and 10μL; Bulb assembly; Spotting holder
0.5, 1, 2, 5, 10 μL
TLC Spotting Capillary Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Microcaps™ TLC Spotting Capillaries > TLC Spotting Capillaries