missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Kimtech™ Absorbent Towels

Use these towels for wiping tasks where low lint or use of chemicals are required. Kimtech™ Absorbent Towels are engineered for critical applications involving solvents, inks and paints.

Brand:  Kimberly-Clark Professional™ 7506

206.38 EUR valid until 2025-12-31
Use promo code "24298" to get your promotional price.


Product Code. 10144914

  • 835.00 € / Pack of 16
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Folded wipers are wrapped in a splash-proof, water-repellent polybag for immediate access to hygienically protected wipers in your work area.
  • Kimtech™ offers a range of reliable, high-performance wipers, each suited to a specific task in controlled areas like transport, healthcare and manufacturing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Absorbent Towel
48 cm
38 cm
White
Folded
Non-sterile
18.9 in.
15 in.
Paper
48 x 38 cm
Wiping tasks where low lint or use of chemicals are required Engineered for critical applications involving solvents, inks and paints
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark Professional™ Kimtech™ Absorbent Towels > Color: White, Dimensions (L x W): 48 x 38 cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.