Learn More
L-Alanyl-L-glutamine, 99%, Thermo Scientific Chemicals
Description
A substitute for glutamine in mammalian cell culture media; stable to heat-sterilization.L-Alanyl-L-glutamine is used as a component in pharmaceuticals, cosmetics and food products. It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency. It provides nutritional requirements, improves intestinal health and enhances the immune system. It is also involved in various clinical studies such as cancer treatment, radiation and chemotherapy. It finds application in biotechnology as a component for mammalian cell culture medium. It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• L-Alanyl-L-glutamine is a synthetic dipeptide that degrades to L-glutamine
Application
• L-Alanyl-L-glutamine can be used as a substitute for glutamine in mammalian cell culture media and is stable to heat-sterilization
• It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency
• It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release
• When this dipeptide is added to cultures of antibody-producing CHO cells in vitro, it reduces apoptosis and promotes antibody production

Specifications
Specifications
Melting Point | 222°C |
Boiling Point | 615°C |
Color | White |
Solubility Information | Soluble in water. |
IUPAC Name | (2S)-5-amino-2-[[(2S)-2-aminopropanoyl]amino]-5-oxopentanoic acid |
Quantity | 100 g |
Flash Point | >110°C (230°F) |
Formula Weight | 217.22 |
Percent Purity | 99% |
Physical Form | Powder |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y11K025 | Certificate of Analysis | 19/11/2024 | J66996.06,J66996.22,J66996.14 |
X25K039 | Certificate of Analysis | 08/11/2024 | J66996.06,J66996.22,J66996.14 |
W07K016 | Certificate of Analysis | 23/09/2024 | J66996.06,J66996.22,J66996.14 |
S27K021 | Certificate of Analysis | 09/07/2024 | J66996.06,J66996.22,J66996.14 |
P01K007 | Certificate of Analysis | 08/03/2024 | J66996.06,J66996.22,J66996.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.