Learn More
Thermo Scientific Chemicals L-(+)-Asparagine, 99%
Description
L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.
Solubility
Soluble in acids, bases. Insoluble in methanol, ethanol, ether and benzene.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 234°C to 236°C (decomposition) |
Beilstein | 1723527 |
Merck Index | 14,837 |
Solubility Information | Soluble in acids,bases. Insoluble in methanol,ethanol,ether and benzene. |
Quantity | 500 g |
Formula Weight | 132.12 |
Sensitivity | Hygroscopic |
Percent Purity | 99% |
Chemical Name or Material | L-(+)-Asparagine |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q23K034 | Certificate of Analysis | 11/06/2024 | B21473.22,B21473.36,B21473.0E |
R22J022 | Certificate of Analysis | 27/06/2023 | B21473.22,B21473,B21473.36,B21473.0E |
A0429707 | Certificate of Analysis | 08/11/2022 | B21473.22,B21473.36,B21473.0E |
A0437909 | Certificate of Analysis | 03/11/2022 | B21473.22,B21473.36,B21473.0E |
U04I011 | Certificate of Analysis | 11/08/2022 | B21473.22,B21473.36,B21473.0E |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.