Learn More
L-Cystine, MedChemExpress
MedChemExpress L-Cystine is an amino acid and intracellular thiol, which plays a critical role in the regulation of cellular processes.
Brand: Medchem Express HY-N0394

Description
L-Cystine is an amino acid and intracellular thiol, which plays a critical role in the regulation of cellular processes.Specifications
L-Cystine | |
White | |
C6H12N2O4S2 | |
0.1 M NaOH : 5 mg/mL (20.81 mM; ultrasonic and adjust pH to 9 with NaOH) ∣0.1 M HCL : 2.5 mg/mL (10.40 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) ∣H2O : < 0.1 mg/mL (insoluble) | |
240.3 | |
97.0% | |
Powder |
56-89-3 | |
Research | |
1 g | |
N[C@@H](CSSC[C@@H](C(O)=O)N)C(O)=O | |
240.3 | |
Research | |
Metabolism-protein/nucleotide metabolism |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only