missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mannitol Salt Agar (Dehydrated)

Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens with this highly selective medium.

Brand:  Thermo Scientific™ CM0085B

108.04 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.


Product Code. 10096012

  • 139.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens with Thermo Scientific™ Oxoid™ Mannitol Salt Agar (Dehydrated). Due to high concentrations of salt in the medium, most unwanted bacteria are inhibited with the exception of some halophilic marine organisms. Presumptive coagulase-positive staphylococci produce colonies surrounded by bright yellow zones while non-pathogenic staphylococci produce colonies with reddish purple zones.

Staphylococcus aureus is a coagulase-positive, catalase-positive, Gram-positive cocci that occur singly or in pairs, tetrads, short chains, and grape-like clusters. S. aureus can produce a variety of syndromes with clinical manifestations, including skin and soft tissue infections, empyema, bloodstream infections, pneumonia, osteomyelitis, septic arthritis, endocarditis, sepsis, and meningitis.

Clearly and easily differentiate Staphylococcus aureus and Staphylococcus epidermidis while most other bacteria is inhibited by the high salt concentration.

  • Highly selective: due to high concentrations of salt in the medium.
  • Easy to differentiate: Staphylococcus aureus forms yellow colonies with yellow halo while Staphylococcus epidermis forms pink colonies with medium turning pink.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mannitol Salt Agar
Agar
For 4.5L medium
Powder
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6246165 Certificate TypeCertificate of Analysis Date13/04/2025 Catalog NumberCM0085B
Lot Number6233016 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberCM0085B
Lot Number6216170 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberCM0085B
Lot Number6206095 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog NumberCM0085B
Lot Number6168656 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberCM0085B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Mannitol Salt Agar (Dehydrated) > 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only