missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ PMMA Workstation Storage
Manufactured from PMMA (acrylic glass.) BRAND™ PMMA Workstation Storage is ideal for storing and dispensing small laboratory items like pipet tips, microcentrifuge tubes, and Pasteur pipets.
Brand: Brand™ 131900
257.75 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from PMMA (acrylic glass)
- White and clear. Available in two sizes.
- Ideal for storing and dispensing small laboratory items like pipet tips, microcentrifuge tubes, and Pasteur pipets.
- Removable top cover with hinged front cover.

Specifications
Workstation Storage | |
(LxWxH) 165 x 152 x 355mm | |
Storing and dispensing small laboratory items like pipet tips, microcentrifuge tubes, and Pasteur pipets | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brand™ PMMA Workstation Storage > Length: 165mm
Spot an opportunity for improvement?Share a Content Correction