missing translation for 'onlineSavingsMsg'
Learn More
L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals Product Code.: 15845288

L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals

Product Code. 15845288 Shop All Thermo Scientific Products
1 kg, Plastic Bottle
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 15845288

Brand: Thermo Scientific J16215.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-VKHMYHEASA-N
Synonym l-glutamic acid, glutamic acid, 2 s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidexShow MoreShow Less
PubChem CID 33032
ChEBI CHEBI:16015
IUPAC Name (2 S)-2-aminopentanedioic acid
SMILES C(CC(=O)O)C(C(=O)O)N

Specifications

Melting Point 189°C to 192°C (decomposition)
Packaging Plastic Bottle
Linear Formula HOOCCH2CH2CH(NH2)COOH
Beilstein 1723801
Density 1.525
Quantity 1 kg
Formula Weight 147.13
Percent Purity 98.5 to 101.5%
Specific Gravity 1.525
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Glutamic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number242295 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ16215.A7,J16215.C4,J16215.A1
Lot Number241204 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ16215.A7,J16215.C4,J16215.A1
Lot Number200313 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ16215.A7,J16215.C4,J16215.A1
Lot Number234017 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberJ16215.A7,J16215.C4,J16215.A1
Lot Number232176 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog NumberJ16215.A7,J16215.C4,J16215.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.