missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- L-Leucine is an essential amino acid that has an important role in the growth and repair of muscle and bone tissue
- It functions to preserve muscle glycogen and helps to prevent the breakdown of muscle proteins after trauma or severe stress
- It is also involved in hemoglobin formation and metabolic functions. This amino acid cannot be synthesized to meet the body's needs from simpler molecules, and therefore must be provided in the diet.
- L-Leucine stimulates protein synthesis by the initiation of mRNA translation
Applications
- L-Leucine is used in the biosynthesis of proteins
- In vivo studies show that L-Leucine can be used when gene defects lead to an organism's inability to metabolize the amino acid phenylalanine
- This compound also contributes to the regulation of blood-sugar levels, growth hormone production, and wound healing
- In research experiments this compound has been used as an activator of the mammalian target of rapamycin (mTOR)

Specifications
Specifications
Melting Point | 286.0°C to 288.0°C |
Specific Rotation | + 15.40 |
Solubility Information | Solubility in water: 22.4g/L (20°C). Other solubilities: 10.9g/L acetic acid, slightly soluble in alcohol, insoluble in ether |
Sublimation Point | 145.0 °C |
Specific Rotation Condition | + 15.40 (20.00°C c=4, 6N HCl) |
Quantity | 500 g |
Formula Weight | 131.17 |
Chemical Name or Material | L-Leucine |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
B0154401 | Certificate of Analysis | 11/06/2024 | 125125000,125121000,125120025 |
B0150359 | Certificate of Analysis | 14/11/2023 | 125125000,125121000,125120025 |
B0150483 | Certificate of Analysis | 14/11/2023 | 125125000,125121000,125120025 |
B0149340 | Certificate of Analysis | 14/11/2023 | 125125000,125121000,125120025 |
B0147174 | Certificate of Analysis | 20/10/2023 | 125125000,125121000,125120025 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals L-Leucine, 99% >
Spot an opportunity for improvement?Share a Content Correction