missing translation for 'onlineSavingsMsg'
Learn More
L(+)-Leucinol, 98%, Thermo Scientific Chemicals Product Code.: 10330994

L(+)-Leucinol, 98%, Thermo Scientific Chemicals

Product Code. 10330994
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10330994

Brand: Thermo Scientific Acros 179970250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7533-40-6
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00063676
InChI Key VPSSPAXIFBTOHY-LURJTMIESA-N
Synonym l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanolShow MoreShow Less
PubChem CID 111307
IUPAC Name (2S)-2-amino-4-methylpentan-1-ol
SMILES CC(C)C[C@H](N)CO

Specifications

Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
Refractive Index 1.4490 to 1.4530 (20°C, 589nm)
Specific Rotation +3° to +5° (20°C, 589nm) (c=9, ethanol)
Density 0.91g/mL
Specific Rotation Condition +4° (20°C 589nm,c=9,ethanol)
Quantity 25 g
Physical Form Liquid
Assay Percent Range 98%
Linear Formula (CH3)2CHCH2CH(NH2)CH2OH
Beilstein 04, 298
Formula Weight 117.19
Percent Purity ≥97.5%
Chemical Name or Material L(+)-Leucinol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0433228 Certificate TypeCertificate of Analysis Date06/09/2023 Catalog Number179970250,179970050
Lot NumberA0390134 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number179970250,179970050
Lot NumberA0410195 Certificate TypeCertificate of Analysis Date30/07/2021 Catalog Number179970250,179970050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L(+)-Leucinol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.