missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Methionine, 98+% Product Code.: 11444083

Thermo Scientific Chemicals L-Methionine, 98+%

Product Code. 11444083
2500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11444083

Brand: Thermo Scientific Chemicals A10318.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Methionine (CAS # 63-68-3) is an essential amino acid that contains sulphur. L-Methionine is involved in cellular detoxification in several species.

L-Methionine iserves as a chelating agent for heavy metals, as a flavoring agent and nutritional supplement for foods. It acts as a feed additive, vegetable oil enrichment and as a single cell protein. In addition to this, it is used as a hepatoprotectant and also acts as a lipotropic agent and prevents excess fat buildup in the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Methionine is an essential amino acid required for tissue repair and maintenance

• L-Methionine affects absorption of selenium and zinc

• L-Methionine can assist cellular detoxification by chelating lead and mercury

• L-Methionine exhibitis properties associated with anti-aging and can also prevent the buildup of fat in the liver

Application

• L-Methionine is appropriate for use in cell culture and biochemistry laboratory applications

• It is useful for the laboratory study of methionine pathways in the cell relating to aging, degeneration, and metabolism

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilaninShow MoreShow Less
PubChem CID 6137
ChEBI CHEBI:16643
IUPAC Name (2S)-2-amino-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point ∼280°C (decomposition)
Beilstein 1722294
Merck Index 14,5975
Solubility Information Soluble in water and hydrochloride. Slightly soluble in acetic acid. Insoluble in ethanol,benzene,acetone and ether.
Quantity 2500 g
Formula Weight 149.21
Optical Rotation +23° (c=2 in 2N HCl)
Percent Purity ≥98%
Chemical Name or Material L-Methionine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM27L023 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA10318.0E,A10318.22,A10318.36
Lot NumberZ16K029 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberA10318.0E,A10318.22,A10318.36
Lot NumberW17K001 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog NumberA10318.0E,A10318.22,A10318.36
Lot NumberT22K039 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog NumberA10318.0E,A10318.22,A10318.36
Lot NumberR20K027 Certificate TypeCertificate of Analysis Date24/05/2024 Catalog NumberA10318.0E,A10318.22,A10318.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Methionine, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.