missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Methionine sulfone, 98+% Product Code.: 11428886

Thermo Scientific Chemicals L-Methionine sulfone, 98+%

Product Code. 11428886
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11428886

Brand: Thermo Scientific Chemicals A17027.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7314-32-1
Molecular Formula C5H11NO4S
Molecular Weight (g/mol) 181.21
MDL Number MFCD00066020
InChI Key UCUNFLYVYCGDHP-BYPYZUCNSA-N
Synonym l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+%Show MoreShow Less
PubChem CID 445282
ChEBI CHEBI:21363
IUPAC Name (2S)-2-amino-4-methylsulfonylbutanoic acid
SMILES CS(=O)(=O)CC[C@H](N)C(O)=O

Specifications

Melting Point 254°C (decomposition)
Beilstein 1725510
Quantity 1 g
Formula Weight 181.21
Optical Rotation +11.6° (c=0.69 in Water)
Percent Purity ≥99%
Chemical Name or Material L-Methionine sulfone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252204 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog NumberA17027.03,A17027.06
Lot Number10239440 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17027.03,A17027.06
Lot Number10228725 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17027.03,A17027.06
Lot NumberJ9560A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17027.03,A17027.06
Lot Number10215774 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17027.03,A17027.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Methionine sulfone, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.