missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L(-)-Proline, 99+% Product Code.: 10205490

Thermo Scientific Chemicals L(-)-Proline, 99+%

Product Code. 10205490
100 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10205490

Brand: Thermo Scientific Chemicals 157621000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Proline CAS 147-85-3, (synonym: (S)-Pyrrolidine-2-carboxylic acid) is a nonessential amino acid and building block of proteins that has applications such as acting as catalyst in chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• L-Proline is an amino acid that is found as a component of various proteins.

Application
• L-Proline is used as asymmetric catalyst in organic synthesis and asymmetric aldol cyclization
• It is also used along with ninhydrin in chromatography
• It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes
• It is used in pharmaceutical and biotechnological applications due to its osmoprotactant property
• It is precursor of hydroxyproline in collage and involved in proper functioning of joint and tendons

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
Synonym l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-ohShow MoreShow Less
PubChem CID 145742
ChEBI CHEBI:17203
SMILES OC(=O)C1CCCN1

Specifications

Ammonium 200ppm max.
Arsenic (As) 1ppm max.
pH 5.5 to 7 (2.5% soln. at 25°C)
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99+%
Loss on Drying 0.3% max. (105°C, 3 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Glass Bottle
Beilstein 22, 2
Specific Rotation -84.5° to -86.8° (20°C, 589nm) (c=4, H2O)
Fieser 06,492; 08,421; 09,393; 10,331; 11,446; 13,292
Sulfate 200ppm max.
Merck Index 15, 7895
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Density 1.3500g/mL
IUPAC Name (2S)-pyrrolidine-2-carboxylic acid
Specific Rotation Condition − 85.50 (20.00°C c=4,H2O)
Quantity 100 g
Formula Weight 115.13
Percent Purity 99.0 to 101.0%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(-)-Proline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0449171 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number157620050,157621000,157620250,157625000
Lot NumberA0466037 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog Number157620050,157621000,157620250,157625000
Lot NumberA0460416 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number157620050,157621000,157620250,157625000
Lot NumberA0449171 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number157620050,157621000,157620250,157625000
Lot NumberA0448151 Certificate TypeCertificate of Analysis Date16/10/2023 Catalog Number157620050,157621000,157620250,157625000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L(-)-Proline, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.