missing translation for 'onlineSavingsMsg'
Learn More

Kartell L-Shape Bacteriology Loops

Disposable L-shaped sterile polystyrene spreaders for bacteriology with special 100° angling for easier use.

Brand:  Kartell 0264700

Product Code. 18037741

  • 379.00 € / Pack of 1000
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Especially handy for dispersing liquids across agar culture plates
  • Available single-wrapped
  • Intended to be used to transfer and spread inoculum from a clinical specimen and/or culture isolate into a culture medium (e.g.agar or broth) for subsequent in vitro diagnostic processing and/or testing
Specifications

Specifications

1.28 in.
0.197 in.
5.75 in.
White
Sterile
32.5 mm
5 mm
146 mm
Cell Spreader
Polystyrene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell L-Shape Bacteriology Loops > Color: White

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.