missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ L Shaped Spreaders
Features smooth edges which avoids tearing and digging into agar
Brand: Microspec™ E10
Description
- High Impact Polystyrene construction
- Full length to avoid contamination
- Disposable
- Sterile, aseptically manufactured
- Ideal for bacterial counts
Specifications
Single-use | |
5.5 in. | |
Microseal Pouch | |
High-impact Polystyrene | |
Aseptically Manufactured |
Blue | |
140 mm | |
500 | |
Inoculation Product |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Microspec™ L Shaped Spreaders > No. per Case: 500
Spot an opportunity for improvement?Share a Content Correction