Learn More
Thermo Scientific Chemicals L-Tyrosine, +99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• L-Tyrosine is the levorotatory isomer of the aromatic amino acid tyrosine, synthesized in vivo from L-phenylalanine
• It is available as a crystalline, off-white powder, stable at room temperature, and soluble in hydrochloric acid
Applications
• In vivo L-tyrosine is involved in protein synthesis and is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catecholamines.
• Used as a supplement, L-tyrosine has been shown to increase levels of the neurotransmitters, dopamine, adrenaline, and norepinephrine in the brain
• It is considered an essential dietary supplement for phenylketonuria patients who lack the enzyme phenylalanine hydroxylase, and hence cannot convert phenylalanine into tyrosine
• L-tyrosine plays a vital role as an electron-carrier in the process of photosynthesis
• Due to its role in the synthesis of catecholamines, L-tyrosine is also used in research to study its impact on stress

Specifications
Specifications
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Loss on Drying | 0.5% max. (1 g, 105°C) |
Packaging | Plastic Bottle |
Linear Formula | 4-(HO)C6H4CH2CH(NH2)CO2H |
Beilstein | 14, 605 |
Specific Rotation | -9.8° to -12.3° (20°C, 589nm) (c=5, 1N HCl) |
Merck Index | 15, 10020 |
Sulfated Ash | 0.1% max. |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0386882 | Certificate of Analysis | 31/01/2025 | 140641000,140640010,140640025,140640050 |
A0433917 | Certificate of Analysis | 25/03/2024 | 140640010,140641000,140640025,140640050 |
A0428187 | Certificate of Analysis | 21/09/2023 | 140640010,140641000,140640025,140640050 |
A0450548 | Certificate of Analysis | 02/05/2023 | 140640010,140641000,140640025,140640050 |
A0448392 | Certificate of Analysis | 14/12/2022 | 140640010,140641000,140640025,140640050 |
Safety and Handling

- L-Tyrosine
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.