Learn More
Labconco™ Stainless Steel Laboratory Cart
Holds a variety of lab supplies, equipment and containers
Brand: Labconco™ 8019000
Description
- Load capacity: 400 lb. (181kg)
- Two removable stainless-steel pans are 2 in. deep (5cm) on three sides; one open side for easy transfer from countertop to cart
- Durable, welded frame with glacier white, corrosion-resistant, baked-on powder coating
- 4 in. dia. (10cm) hard rubber casters stabilize movement over rough, uneven floors; two casters have toe locks
- Lifetime warranty
Order Info
Shipped fully assembled. Weight: 50 lb. (23kg).
Services
Lifetime guarantee.
Specifications
34.88 x 19.0 x 36.38 in. (88.6 x 48.3 x 92.4 cm) | |
Two removable polished Type 304 stainless steel pans; 4 in. diameter casters, two with toe locks; Extruded rubber handle grips; Glacier white, powder-coated 18 gauge welded tubular steel frame; ID tag | |
400 lb. | |
Steel |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.