Learn More
Buerkle™ Polyethylene Square All-Purpose Boxes
Buerkle™ Polyethylene Square All-Purpose Boxes are for storing, packing, taking samples, sampling, and transport of pastes, granules, powders, soil samples and frozen goods.
3.90 € - 14.20 €
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
10096471
|
Buerkle™
6250-0500 |
500 mL | 103 x 103 x 64 mm |
3.90 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10475582
|
Buerkle™
6250-1000 |
1000 mL | 103 x 103 x 127 mm |
5.03 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10659012
|
Buerkle™
6250-1001 |
1000 mL | 208 x 103 x 64 mm |
7.10 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10310642
|
Buerkle™
6250-2000 |
2000 mL | 208 x 208 x 64 mm |
11.28 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10023373
|
Buerkle™
6250-2300 |
2000 mL | 255 x 103 x 94 mm |
10.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10390682
|
Buerkle™
6250-3300 |
3200 mL | 208 x 208 x 94 mm |
14.20 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10770583
|
Buerkle™
6250-1500 |
1500 mL | 208 x 103 x 94 mm |
9.55 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Frozen goods from −40° up to briefly +95°C
- Ideal use of space, can be stacked
- Comes complete with press-in lid
- Transparent in color
- Can be deep frozen
- Smooth surface, no crevices
Specifications
Transparent | |
Polyethylene | |
Square |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Buerkle™ Polyethylene Square All-Purpose Boxes