missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts Legionella Positive Control Reagent
Microgen Bioproducts Legionella Positive Control reagent
Brand: Novacyt - Microgen Bioproducts M45C
This item is not returnable.
View return policy
Description
- Individual L.pneumophila serogroups 1, 2-15, and non L.pneumophila species latex and positive control available separately.
Specifications
ISO13485 ISO9001 | |
Yes | |
1 Bottle (1.0 mL) | |
1 | |
The Confirmatory Identification of Legionella Pneumophila and Other Commonly Isolated Legionellasp Ecies Grown on Selective Medi | |
1.0 mL | |
1/Test | |
Bottle in polygrip bag | |
Legionella pneumophila | |
Bacteria |
2°C to 8°C | |
Non-waived | |
Bacterial Colonies | |
Reagent | |
0.85% Isotonic Saline, Disposable Agglutination Slides, Disposable Mixing Sticks, Bacteriological Loops, Legionella Selective Medium, Glass Tubes for Boiling | |
Polyethylene Dropper Bottle | |
Monera | |
1.0 mL | |
1 | |
7 minutes |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novacyt - Microgen Bioproducts Legionella Positive Control Reagent > Legionella Positive Control Reagent
Spot an opportunity for improvement?Share a Content Correction