missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ IP Lysis Buffer

Maximize cell lysate yield, purity and compatibility with this immunoprecipitation-optimized protein extraction reagent.

Brand:  Thermo Scientific™ 87787

118.75 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 11820084

  • 128.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce IP Lysis Buffer is optimized for cell lysate yield, purity and compatibility with immunoprecipitation.

The IP Lysis Buffer is a mammalian whole cell lysis reagent based on a modified RIPA buffer formulation without SDS. This moderate-strength lysis buffer effectively solubilizes cellular proteins but does not liberate genomic DNA or disrupt protein complexes like ordinary RIPA buffer. Pierce IP Lysis Buffer is specially formulated for pulldown and immunoprecipitation assays.

Highlights:

  • Optimized for compatibility with immunoprecipitation and pull-down assays
  • Compatible with protein assays, reporter assays and immunoassay procedures
  • Ready-made formula is effective for extracting cytoplasmic, membrane and nuclear proteins
  • Gentle formulation helps maintain protein complexes for co-immunoprecipitation (co-IP)
  • Does not liberate genomic DNA which can cause high sample viscosity
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
IP Lysis Buffer
Upon receipt store at 4°C.
Liquid
100 mL
Pierce™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA407472 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number87787
Lot NumberZL403858 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number87787
Lot NumberZJ405514 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog Number87787
Lot NumberZH403857 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number87787
Lot NumberZH402864 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog Number87787
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ IP Lysis Buffer > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.