Learn More
Thermo Scientific™ DrySpot™ Legionella Pneumophila Serogroup 2-14 Latex Agglutination Test
Rapidly confirm Legionella pneumophila serogroups 2-14 from culture by latex agglutination.
Brand: Thermo Scientific™ DR0210M
Description
The DrySpot Legionella Latex Test is a latex agglutination test for the identification of Legionella species grown on plate media from patients with suspected legionellosis or from environmental sources. The DrySpot Legionella Latex Test allows separate identification of Legionella pneumophila serogroup 1, serogroups 2–14 and seven other Legionella species that have been implicated in human disease.
Use this dry latex agglutination test for the identification of Legionella pneumophila serogroups 2-14, isolated either from patients with suspected legionellosis or from environmental sources.Blue latex particles sensitised with specific rabbit antibody are dried onto the test card. These react with Legionella pneumophila serogroup 2-14 antigen to form agglutination in the test reaction area.
Blue latex particles sensitised with non-reactive rabbit globulins are dried onto the test card. These provide the control reaction area.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
2°C to 25°C | |
Legionella | |
Legionella Latex Test Kit | |
60 Tests/Kit |
DrySpot Legionella Pneumophila Serogroup 2-14 Latex Test | |
Environmental Source | |
20 reaction cards (3 tests/card), positive control, negative control, phosphate buffered saline, mixing paddles, pouch clips |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6193323 | Certificate of Analysis | 11/03/2025 | DR0210M |
6139990 | Certificate of Analysis | 18/11/2024 | DR0210M |
3806042 | Certificate of Analysis | 10/06/2024 | DR0210M |
3754908 | Certificate of Analysis | 15/02/2024 | DR0210M |
3666800 | Certificate of Analysis | 28/09/2023 | DR0210M |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.