Learn More
Ley Rubber Rubber Flask Adapter
Suitable for use with: Gooch funnels, sintered glass funnels, slit sleeve funnels and porcelain funnels.
Brand:  Fisherbrand CWF-740-D/Z
Product Code. 12378845
  • 32.00 € / Pack of 5
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pliable, molded rubber sleeve to fit the conical part of Gooch crucible adapters and small Buchner funnels and make a vacuum-tight joint with the rim of a filter flask
Specifications
Specifications
50 x 24 x 36mm
Gooch funnels, sintered glass funnels, slit sleeve funnels and porcelain funnels
Flask Adapter
Rubber
5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ley Rubber Rubber Flask Adapter > Size: 50 x 24 x 36mm