Learn More
Biochrom Lightwave 3
The Lightwave 3 is a split beam UV/Vis spectrophotometer that combines ease of use with the flexibility of a 40 mm sample port.
Brand:  Biochrom 80-3007-42
 View more versions of this product
Product Code. 17143628
  • 6835.00 € / Each
Explore more special offers
Description
Description
  • Split Beam (RBC) UV/Vis Spectrophotometer covering 190 to 1100 nm (5 nm bandwidth)
  • USB port for data collection
  • Accepts 10, 20, and 40 mm path length cuvettes
  • Built-in software provides flash scan, fixed wavelength measurements, kinetics, and concentration with comprehensive graphics capability plus the ability to store up to 90 methods
  • Choose to include a built-in serial printer, Bluetooth connectivity, or both
  • Lightweight and stackable for storage
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Biochrom Lightwave 3 > Biochrom Lightwave 3; Includes: Spectrophotometer; Interface: Large Display, Button UI, USB Port, PC via PVC Software; Printer Included: No