missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Live Cell Imaging Solution

The Live Cell Imaging Solution from Molecular Probes™ is a physiological medium developed for live cell imaging applications that has universal utility in cell-based research—for live cell imaging, dye loading, and wash steps.

Brand:  Invitrogen™ A59688DJ

Product Code. 17927162

  • 30.48 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Live Cell Imaging Solution from Molecular Probes™ is a physiological medium developed for live cell imaging applications that has universal utility in cell-based research—for live cell imaging, dye loading, and wash steps. It is an optically clear, physiological solution buffered with HEPES at pH 7.4 that keeps cells healthy for up to 4 hours at ambient atmosphere and temperature. Compared to standard media, Live Cell Imaging Solution provides better clarity, signal-to-noise ratio, and cell viability.

Outstanding Features of Live Cell Imaging Solution:
Physiological and chemical optimization—providing the ideal solution for live cell fluorescence imaging applications
Optical clarity—affording more efficient imaging and minimal background
Extended buffering capacity compared to media—for improved cell viability

Obtain Great Results with a Medium Designed to Meet the Demands of Live Cell Imaging
Live cell imaging can be challenging. Not only must staining be optimized for the particular assay readout, spectral compatibility, and signal-to-noise level, but live cell imaging of dynamic processes and cell physiology requires live observation over time. The choice of medium is particularly important for time-lapse imaging and experiments where cells are exposed to ambient conditions for longer periods. For reliable results, it is essential that cells be healthy and maintained as closely as possible to physiological temperature, pH, oxygen tension, and other conditions. If cells appear unhealthy they should not be used for imaging. Live Cell Imaging Solution has been developed specifically for live cell imaging applications; as a result, it provides image clarity (Fig 1), signal-to-noise ratio (Fig 2), and cell viability that are significantly enhanced compared to standard media. In addition to live cell imaging, the physiological composition of the Live Cell Imaging Solution makes it ideal for other cell-based applications, such as wash steps and dye loading.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Molecular Probes

Store at ≤25°C.

500 mL
Physiological Imaging Buffer
Sustainable packaging
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Live Cell Imaging Solution > 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.