Learn More
Thermo Scientific™ LN2 Low-Level Alarms
Add an extra level of protection to the lab in the event of excessive liquid nitrogen loss during dewar storage with LN2 Low-Level Alarms.
Brand:  Thermo Scientific™ 8130
Product Code. 12774708
  • 1366.80 € / Each
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Add an extra level of protection to the laboratory in the event of excessive liquid nitrogen loss during dewar storage with Thermo Scientific™ LN2 Low-Level Alarms, for use with any standard-sized LN2 container.
Low level alarm (120V) with sensor for any standard-sized LN2 container.
Includes:
Dry contacts for remote alarm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Low-level alarm with dry contacts for connection to telephone dialing system or remote alarm, customer installed
Any Standard-Sized LN2 Container
120 V
LN2 Low-Level Alarm
Alarm
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ LN2 Low-Level Alarms > Forma Ll Alarm