missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
GREENER_CHOICE

LO-tox™ lab thermometer complete with calibration certificate.

Specifications

Material Glass
Width (Metric) 1.2 cm
View More Specs

Products 24
Product Code Brand Temperature Range (Metric) Subdivision Immersion Type Length (Metric) Price Quantity & Availability  
Product Code Brand Temperature Range (Metric) Subdivision Immersion Type Length (Metric) Price Quantity & Availability  
17882988
GREENER_CHOICE
View Documents
Brannan™
44/825/0 + 49/503/0
-1°C to 101°C 0.2°C Total Immersion 40.5 cm N/A
N/A
17843298
GREENER_CHOICE
View Documents
Brannan™
44/824/0 + 49/503/0
-1°C to 101°C 0.2°C Partial Immersion 40.5 cm N/A
N/A
17892978
GREENER_CHOICE
View Documents
Brannan™
44/820/0 + 49/503/0
-1°C to 51°C 0.1°C Partial Immersion 40.5 cm N/A
N/A
17872988
GREENER_CHOICE
View Documents
Brannan™
44/826/8 + 49/503/0
-10°C to +360°C 2°C Partial Immersion 30.5 cm N/A
N/A
17832988
GREENER_CHOICE
View Documents
Brannan™
44/822/0 + 49/503/0
-1°C to 51°C 0.2°C Partial Immersion 40.5 cm N/A
N/A
17802998
GREENER_CHOICE
View Documents
Brannan™
44/803/8 + 49/503/0
-20°C to +150°C 1°C Partial Immersion 30.5 cm N/A
N/A
17893288
GREENER_CHOICE
View Documents
Brannan™
44/804/8 + 49/503/0
-10°C to +260°C 1°C Partial Immersion 40.5 cm N/A
N/A
17833008
View Documents
Brannan™
44/830/7 + 49/503/0
-10°C to +110°C 1°C Total Immersion 15.5 cm N/A
N/A
17853068
View Documents
Brannan™
44/810/9 + 49/503/0
-20°C to +110°C 0.5°C Total Immersion 30.5 cm N/A
N/A
17843068
GREENER_CHOICE
View Documents
Brannan™
44/805/8 + 49/503/0
-10°C to +210°C 1°C Partial Immersion 40.5 cm N/A
N/A
17803078
View Documents
Brannan™
44/802/8 + 49/503/0
-20°C to +110°C 1°C Partial Immersion 30.5 cm N/A
N/A
17863288
GREENER_CHOICE
View Documents
Brannan™
44/810/8 + 49/503/0
-20°C to +110°C 1°C Total Immersion 30.5 cm N/A
N/A
17803298
GREENER_CHOICE
View Documents
Brannan™
44/811/8 + 49/503/0
-20°C to +150°C 1°C Total Immersion 30.5 cm N/A
N/A
17893068
GREENER_CHOICE
View Documents
Brannan™
44/819/0 + 49/503/0
-10°C to +360°C 2°C Total Immersion 30.5 cm N/A
N/A
17813118
GREENER_CHOICE
View Documents
Brannan™
44/809/8 + 49/503/0
-35°C to +50°C 1°C Total Immersion 30.5 cm N/A
N/A
17833088
GREENER_CHOICE
View Documents
Brannan™
44/812/8 + 49/503/0
-10°C to +210°C 1°C Total Immersion 40.5 cm N/A
N/A
17833068
GREENER_CHOICE
View Documents
Brannan™
44/823/0 + 49/503/0
-1°C to 51°C 0.2°C Total Immersion 40.5 cm N/A
N/A
17813078
View Documents
Brannan™
44/802/9 + 49/503/0
-20°C to +110°C 0.5°C Partial Immersion 30.5 cm N/A
N/A
17813298
GREENER_CHOICE
View Documents
Brannan™
44/801/8 + 49/503/0
-35°C to +50°C 0.5°C Partial Immersion 30.5 cm N/A
N/A
17853298
GREENER_CHOICE
View Documents
Brannan™
44/800/8 + 49/503/0
-10°C to +50°C 0.5°C Partial Immersion 30.5 cm N/A
N/A
17883108
GREENER_CHOICE
View Documents
Brannan™
44/808/8 + 49/503/0
-10°C to +50°C 0.5°C Total Immersion 30.5 cm N/A
N/A
17883298
GREENER_CHOICE
View Documents
Brannan™
44/821/0 + 49/503/0
-1°C to 51°C 0.1°C Total Immersion 40.5 cm N/A
N/A
17873068
GREENER_CHOICE
View Documents
Brannan™
44/815/0 + 49/503/0
-10°C to +300°C 1°C Partial Immersion 30.5 cm N/A
N/A
17843078
View Documents
Brannan™
44/850/7 + 49/503/0
-10°C to +50°C 1°C Total Immersion 15.5 cm N/A
N/A
Description

Description

  • LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications

Specifications

Glass
1.2 cm
Videos
SDS
Documents

Documents

Product Certifications