missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Nalgene LOCK TILL PETG MEDIA FL 38-430 FP240
GREENER_CHOICE

Brand:  Thermo Scientific Nalgene 342151-0380

107.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


LOCK TILL PETG MEDIA FL 38-430 FP240

Product Code. 11364455

  • 108.07 € / Pack of 200

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number7450412010 Certificate TypeCertificate of Processing Date31/03/2025 Catalog Number342151-0380
Lot Number7443087010 Certificate TypeCertificate of Processing Date22/01/2025 Catalog Number342151-0380
Lot Number7450412010 Certificate TypeCertificate of Compliance Date31/03/2025 Catalog Number342151-0380
Lot Number7443087010 Certificate TypeCertificate of Compliance Date22/01/2025 Catalog Number342151-0380
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Nalgene LOCK TILL PETG MEDIA FL 38-430 FP240 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.