Learn More
Agilent BioTek LogPhase 600 Product Qualification Package
LogPhase 600 Product Qualification Package is for use with LogPhase 600 Microbiology Reader
Brand: Agilent BioTek 1800547
Alert:
Note:
Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
Description
Specifications
LogPhase 600 Microbiology Reader; Requires Gen5 309 or Higher; Agilent BioTek Microplate Washer and Reader Instruments | |
Product Qualification Package |
Product Suggestions
Customers who viewed this item also viewed.