missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Powder Packs™ for Free Chlorine

Premeasured unit dose DPD reagents for water quality testing

Brand:  Lovibond™ 530100

 View more versions of this product

Product Code. 10426303

  • 29.45 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Premeasured unit dose DPD reagents for water quality testing

  • For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed foil packets eliminate risk of cross-contamination
  • Available for 5, 10, and 25mL sample sizes
  • Extended shelf life of up to 5 years

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Compliance

US EPA-accepted DPD method

Specifications

Specifications

US EPA
Powder
SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
0 to 2 mg/L
0.33 oz.
10 mL
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
100 Pack
DPD Free Chlorine Reagent
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

SDS
Documents

Documents

Brochures

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number20007582 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog Number530100
Lot Number2414013 Certificate TypeCertificate of Analysis Date17/07/2024 Catalog Number530100
Lot Number2409818 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number530100
Lot Number2406307 Certificate TypeCertificate of Analysis Date06/04/2024 Catalog Number530100
Lot Number2324410 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog Number530100
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lovibond™ Powder Packs™ for Free Chlorine > Free Chlorine; 10mL; Pack of 100

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.