missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Low-Density Polyethylene Bags
Store a variety of solids or powders in these LDPE bags. Low Density Polyethylene Bags stand up to your storage needs with a strong bottom seal.
Brand: Cole-Parmer™ 06505-16
This item is not returnable.
View return policy
Description
- Seal with heat sealer or bag ties
- Good for general laboratory storage
- Available in a variety of sizes and thicknesses
Specifications
Low-Density Bag | |
Polyethylene | |
3 mil | |
305 mm |
Clear | |
500 Pack | |
305 x 254mm | |
254 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Low-Density Polyethylene Bags > 305L x 255mmW
Spot an opportunity for improvement?Share a Content Correction