missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles

Buerkle™ Lowering Cable for dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials.

Brand:  Buerkle™ 5309-0550

 View more versions of this product

Product Code. 17280574

  • 687.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Good chemical resistance
  • For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
  • Tensile load max. up to 10kg
Specifications

Specifications

Lowering Cable
Dipping Samplers and Immersion Cylinders
100 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles > 100 m

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.