missing translation for 'onlineSavingsMsg'
Learn More
Lumefantrine, 98%, Thermo Scientific Chemicals Product Code.: 15781759

Lumefantrine, 98%, Thermo Scientific Chemicals

Product Code. 15781759
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15781759

Brand: Thermo Scientific Acros 461110050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82186-77-4
Molecular Formula C30H32Cl3NO
Molecular Weight (g/mol) 528.94
MDL Number MFCD05662268
InChI Key DYLGFOYVTXJFJP-MYYYXRDXNA-N
IUPAC Name 2-(dibutylamino)-1-[(9Z)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]-9H-fluoren-4-yl]ethan-1-ol
SMILES CCCCN(CCCC)CC(O)C1=CC(Cl)=CC2=C1C1=CC=C(Cl)C=C1\C2=C\C1=CC=C(Cl)C=C1

Specifications

Melting Point 136.0°C
Assay Percent Range 97.5 % min. (HPLC)
Quantity 5 g
Formula Weight 528.94
Chemical Name or Material Lumefantrine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460035 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number461110250,461110050
Lot NumberA0451209 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number461110250,461110050
Lot NumberA0374685 Certificate TypeCertificate of Analysis Date15/09/2021 Catalog Number461110250,461110050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lumefantrine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.