missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagicMark™ XP Western Protein Standard

Specifically designed for easy and convenient protein molecular weight estimation directly on Western blots

Brand:  Invitrogen™ LC5603

101.65 EUR valid until 2025-12-31
Use promo code "24372" to get your promotional price.


 View more versions of this product

Product Code. 10610856

  • 112.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: 50 μL of ready-to-use standard mixture is provided in a plastic vial; Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue
This item is not returnable. View return policy

Description

Description

  • MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site
  • The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot
  • The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent)
  • The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use
  • Store at -20°C

Applications

  • Western blotting: detection of the nine unstained bands via the detection method used for the target protein. Compatible with chemiluminescent substrates and fluorescent secondary antibodies (not recommended for antibodies labeled with fluors in the 500–550 nm channel)
  • Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 μL of ready-to-use standard mixture is provided in a plastic vial. Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue.

Store at -20°C.
9
Yes
Unstained
220, 120, 100, 80, 60, 50, 40, 30, 20 kDa
50 μL
Western Blotting, SDS-PAGE
User defined detection system
Protein Ladder
20 to 220 kDa
Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
MagicMark™
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3063115 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberLC5603
Lot Number2999822 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog NumberLC5603
Lot Number2502000 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberLC5603
Lot Number2545407 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberLC5603
Lot Number2545446 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberLC5603
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ MagicMark™ XP Western Protein Standard > 50 μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.