missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagicMark™ XP Western Protein Standard
Specifically designed for easy and convenient protein molecular weight estimation directly on Western blots
112.00 € - 321.00 €
Specifications
Detection Method | User defined detection system |
---|---|
Number of Markers | 9 |
Ready to Load | Yes |
Size Range | 20 to 220 kDa |
Stain Type | Unstained |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10227592
|
Invitrogen™
LC5602 |
250 μL |
321.00 €
250µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10610856
|
Invitrogen™
LC5603 |
50 μL |
112.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site
- The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot
- The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent)
- The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use
- Store at -20°C
Applications
- Western blotting: detection of the nine unstained bands via the detection method used for the target protein. Compatible with chemiluminescent substrates and fluorescent secondary antibodies (not recommended for antibodies labeled with fluors in the 500–550 nm channel)
- Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method
Order Info
Shipping Condition: Dry Ice
Specifications
User defined detection system | |
Yes | |
Unstained | |
MagicMark™ | |
Western Blotting, SDS-PAGE |
9 | |
20 to 220 kDa | |
220, 120, 100, 80, 60, 50, 40, 30, 20 kDa | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ MagicMark™ XP Western Protein Standard