Learn More
Invitrogen™ MagicMark™ XP Western Protein Standard
Specifically designed for easy and convenient protein molecular weight estimation directly on Western blots
Brand: Invitrogen™ LC5602
239.40 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: 250 μL of ready-to-use standard mixture is provided in a plastic vial; Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue
Description
- MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site
- The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot
- The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent)
- The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use
- Store at -20°C
Applications
- Western blotting: detection of the nine unstained bands via the detection method used for the target protein. Compatible with chemiluminescent substrates and fluorescent secondary antibodies (not recommended for antibodies labeled with fluors in the 500–550 nm channel)
- Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method
Order Info
Shipping Condition: Dry Ice

Specifications
250 μL of ready-to-use standard mixture is provided in a plastic vial. Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue. Store at -20°C. |
|
9 | |
Yes | |
Unstained | |
220, 120, 100, 80, 60, 50, 40, 30, 20 kDa | |
250 μL | |
Western Blotting, SDS-PAGE |
User defined detection system | |
Protein Ladder | |
20 to 220 kDa | |
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels | |
MagicMark™ | |
Dry Ice |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3068506 | Certificate of Analysis | 31/01/2025 | LC5602,LC5602_LITL18 PROMO |
2455212 | Certificate of Analysis | 31/01/2025 | LC5602,LC5602_LITL18 PROMO |
3009454 | Certificate of Analysis | 31/01/2025 | LC5602,LC5602_LITL18 PROMO |
3032414 | Certificate of Analysis | 30/01/2025 | LC5602,LC5602_LITL18 PROMO |
2470188 | Certificate of Analysis | 29/01/2025 | LC5602,LC5602_LITL18 PROMO |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.