Learn More
Manganese(II) acetate, 98+%, anhydrous, Thermo Scientific Chemicals
30.25 € - 271.35 €
Chemical Identifiers
CAS | 638-38-0 |
---|---|
Molecular Formula | C4H6MnO4 |
Molecular Weight (g/mol) | 173.03 |
MDL Number | MFCD00013039 |
InChI Key | UOGMEBQRZBEZQT-UHFFFAOYSA-L |
Synonym | manganese ii acetate, manganese acetate, manganous acetate, diacetylmanganese, manganese diacetate, manganese di acetate, manganese 2+ acetate, octan manganaty czech, acetic acid, manganese 2+ salt, unii-0v6e9q2i0y |
PubChem CID | 12525 |
IUPAC Name | manganese(2+);diacetate |
SMILES | [Mn++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10540374
|
Thermo Scientific Acros
389140050 |
5 g | Glass bottle |
30.25 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10782264
|
Thermo Scientific Acros
389140250 |
25 g | Glass bottle |
103.52 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10238763
|
Thermo Scientific Acros
389141000 |
100 g | Glass bottle |
271.35 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
638-38-0 | |
173.03 | |
UOGMEBQRZBEZQT-UHFFFAOYSA-L | |
12525 | |
[Mn++].CC([O-])=O.CC([O-])=O |
C4H6MnO4 | |
MFCD00013039 | |
manganese ii acetate, manganese acetate, manganous acetate, diacetylmanganese, manganese diacetate, manganese di acetate, manganese 2+ acetate, octan manganaty czech, acetic acid, manganese 2+ salt, unii-0v6e9q2i0y | |
manganese(2+);diacetate |
Specifications
638-38-0 | |
Powder | |
C4H6MnO4 | |
MFCD00013039 | |
manganese ii acetate, manganese acetate, manganous acetate, diacetylmanganese, manganese diacetate, manganese di acetate, manganese 2+ acetate, octan manganaty czech, acetic acid, manganese 2+ salt, unii-0v6e9q2i0y | |
UOGMEBQRZBEZQT-UHFFFAOYSA-L | |
manganese(2+);diacetate | |
12525 | |
≥98% | |
Manganese(II) acetate |
Beige-Gray to Brown | |
98% min. (Complexometry) | |
Mn(OC(O)CH3)2 | |
15, 5789 | |
Solubility in water: very soluble | |
[Mn++].CC([O-])=O.CC([O-])=O | |
173.03 | |
173.03 | |
Authentic |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 211-334-3
- Manganese(II) acetate
RUO – Research Use Only