missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Maxi Horizontal Gel System

Includes 200 x 100mm and 200 x 200mm gel trays. Fisherbrand™ Maxi Horizontal Gel System is primaryily designed for separating high numbers of samples from PCR or cloning.

Brand:  Fisherbrand™ FMSMAXIDUO

Product Code. 11843303

  • 815.00 â‚¬ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Supplied with 200 x 100mm and 200 x 200mm gel trays
  • 200 x 250mm is also available
  • Run up to 550 samples
  • Low buffer volumes
  • Ideal for extended separations
  • Injection molded
  • Primarily designed for separating high numbers of samples from PCR or cloning.
Specifications

Specifications

Up to 550 Samples
200 x 250 mm Gel Trays
200 x 100mm and 200 x 200mm gel trays
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisherbrand and Fisher BioReagents Focus on electrophoresis

    • Fisherbrand - Meet the Family
      Fisher Scientific's private label product line, Fisherbrand™, offers customers a broad selection of laboratory supplies & consumables, covering a range of applications such as pH & electrochemistry, stirring & mixing, centrifugation, liquid handling,
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Fisherbrand Meet the Family

      SDS
      Documents

      Documents

      Certificates
      View accessory products
      Special Offers

      Special Offers

      Product Content Correction

      Your input is important to us. Please complete this form to provide feedback related to the content on this product.

      Product Title
      Fisherbrand™ Maxi Horizontal Gel System > Length: 230mm

      By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

      Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.