Learn More
Spilfyter™ Mercury Spill Cleanup Kit
Mercury Spill Cleanup Kit - Self contained kits provide a safe and efficient way to clean up small mercury spills.
Brand: Spilfyter™ 86464-00
Description
- Safely decontaminate, clean and dispose of hazardous spills
Specifications
Mercury Spill Cleanup Kit | |
Small Mercury Spillage | |
Amalgamation powder, 270 g; Indicator powder, 250 g; Vapour suppressor shaker bottle; Aspirator bottle; Waste collection bottle; Mixing tub with spatula; Chemical sponge; One pair of nitrile gloves; One disposal bag; One wisk pan |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- MERCSORB Mercury Spill Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands, arms, and exposed areas thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see section 4 on this label).
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362-Take off contaminated clothing.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sulphur, copper iodide, zinc, citric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.