missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécification
Spécification
Color | Colorless |
Density | 1.0800g/mL |
Boiling Point | 166.0°C to 169.0°C |
Flash Point | 60°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | HSCH2CH2CO2CH3 |
Refractive Index | 1.4620 to 1.4640 |
Quantity | 100 g |
Afficher plus |
Suggestions de produits
Customers who viewed this item also viewed
Viewing 1-5 of 10
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Sécurité et traitement


Identifiant produit
- Methyl 3-mercaptopropionate
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Methyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu