missing translation for 'onlineSavingsMsg'
Learn More
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, 97%, Thermo Scientific Chemicals Product Code.: 11415453

Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, 97%, Thermo Scientific Chemicals

Product Code. 11415453
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11415453

Brand: Thermo Scientific Alfa Aesar A13418.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.

Solubility
It is soluble in water, acetone, benzene, ethyl ether

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3162-96-7
Molecular Formula C14H18O6
Molecular Weight (g/mol) 282.292
MDL Number MFCD00006819
InChI Key VVSWDMJYIDBTMV-BTZLDLHRSA-N
Synonym 4ar,6s,7r,8r,8as-6-methoxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidene-alpha-d-glucopyranoside, methyl 4,6-o-benzylidene-a-d-glucopyranoside, 4ar,6s,7r,8r,8as-6-methoxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidenehexopyranoside #, zlchem 511, methyl 4,6-o-benzylidene-, a-d-glucopyranoside, methyl 4,6-o-phenylmethylene-, glucopyranoside, methyl 4,6-o-benzylidene-, .alpha.-d, .alpha.-d-glucopyranoside, methyl 4,6-o-phenylmethyleneShow MoreShow Less
PubChem CID 11822086
IUPAC Name (4aR,6S,7R,8R,8aS)-6-methoxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol
SMILES COC1C(C(C2C(O1)COC(O2)C3=CC=CC=C3)O)O

Specifications

Melting Point 163°C to 166°C
Quantity 5 g
Beilstein 1291458
Sensitivity Hygroscopic
Solubility Information It is soluble in water,acetone,benzene,ethyl ether
Optical Rotation +110° (c=2 in Chloroform)
Formula Weight 282.29
Percent Purity 97%
Chemical Name or Material Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241103 Certificate TypeCertificate of Analysis Date29/06/2022 Catalog NumberA13418.22,A13418.14,A13418.06
Lot Number10232044 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13418.22,A13418.14,A13418.06
Lot Number10199762 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13418.22,A13418.14,A13418.06
Lot NumberS16C012 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13418.22,A13418.14,A13418.06
Lot Number10180012 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13418.22,A13418.14,A13418.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.