Learn More
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, 97%, Thermo Scientific Chemicals
Description
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.
Solubility
It is soluble in water, acetone, benzene, ethyl ether
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 163°C to 166°C |
Quantity | 25 g |
Beilstein | 1291458 |
Sensitivity | Hygroscopic |
Solubility Information | It is soluble in water,acetone,benzene,ethyl ether |
Optical Rotation | +110° (c=2 in Chloroform) |
Formula Weight | 282.29 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10241103 | Certificate of Analysis | 29/06/2022 | A13418.22,A13418.14,A13418.06 |
10232044 | Certificate of Analysis | 22/11/2021 | A13418.22,A13418.14,A13418.06 |
10199762 | Certificate of Analysis | 07/08/2020 | A13418.22,A13418.14,A13418.06 |
S16C012 | Certificate of Analysis | 07/08/2020 | A13418.22,A13418.14,A13418.06 |
10180012 | Certificate of Analysis | 07/08/2020 | A13418.22,A13418.14,A13418.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.