missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-oxotetrahydrothiophene-3-carboxylate, Technical Grade, Thermo Scientific™

48.30 € - 585.00 €

Chemical Identifiers

CAS 2689-68-1
Molecular Formula C6H8O3S
Molecular Weight (g/mol) 160.187
InChI Key LEAKUJFYXNILRB-UHFFFAOYSA-N
Synonym methyl 4-oxotetrahydrothiophene-3-carboxylate, 4-carbomethoxytetrahydro-3-thiophenone, 4-carbomethoxy-3-thiophanone, 3-thiophenecarboxylic acid, tetrahydro-4-oxo-, methyl ester, methyl tetrahydro-4-oxo-3-thiophenecarboxylate, methyl 3-oxotetrahydrothiophene-4-carboxylate, 4-carbomethoxytetrahydro-3-thi ophenone, methyl 4-oxo-tetrahydrothiophene-3-carboxylate, methyl 4-oxo-2,3,5-trihydrothiophene-3-carboxylate, methyl tetrahydro-4-oxo-3-thenoate
PubChem CID 98424
IUPAC Name methyl 4-oxothiolane-3-carboxylate
SMILES COC(=O)C1CSCC1=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10531872
View Documents
Thermo Scientific Maybridge
KM00409DA
1 g Amber Glass Bottle
48.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
10752293
View Documents
Thermo Scientific Maybridge
KM00409EA
10 g Amber Glass Bottle
325.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
10622885
View Documents
Thermo Scientific Maybridge
KM00409EB
25 g Amber Glass Bottle
585.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

2689-68-1
160.187
methyl 4-oxotetrahydrothiophene-3-carboxylate, 4-carbomethoxytetrahydro-3-thiophenone, 4-carbomethoxy-3-thiophanone, 3-thiophenecarboxylic acid, tetrahydro-4-oxo-, methyl ester, methyl tetrahydro-4-oxo-3-thiophenecarboxylate, methyl 3-oxotetrahydrothiophene-4-carboxylate, 4-carbomethoxytetrahydro-3-thi ophenone, methyl 4-oxo-tetrahydrothiophene-3-carboxylate, methyl 4-oxo-2,3,5-trihydrothiophene-3-carboxylate, methyl tetrahydro-4-oxo-3-thenoate
methyl 4-oxothiolane-3-carboxylate
C6H8O3S
LEAKUJFYXNILRB-UHFFFAOYSA-N
98424
COC(=O)C1CSCC1=O

Specifications

2689-68-1
methyl 4-oxotetrahydrothiophene-3-carboxylate, 4-carbomethoxytetrahydro-3-thiophenone, 4-carbomethoxy-3-thiophanone, 3-thiophenecarboxylic acid, tetrahydro-4-oxo-, methyl ester, methyl tetrahydro-4-oxo-3-thiophenecarboxylate, methyl 3-oxotetrahydrothiophene-4-carboxylate, 4-carbomethoxytetrahydro-3-thi ophenone, methyl 4-oxo-tetrahydrothiophene-3-carboxylate, methyl 4-oxo-2,3,5-trihydrothiophene-3-carboxylate, methyl tetrahydro-4-oxo-3-thenoate
COC(=O)C1CSCC1=O
160.187
160.19
Technical
C6H8O3S
LEAKUJFYXNILRB-UHFFFAOYSA-N
methyl 4-oxothiolane-3-carboxylate
98424
Tech.
Methyl 4-oxotetrahydrothiophene-3-carboxylate
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.